SP Bel-Art Powder Funnels, Polypropylene, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
H14660-0065 H14660-0080 H14660-0100 H14660-0150 H14660-0150
47750-820PK 105.41 CAD
47750-820 47750-822 47750-824 10033-460 490012-376
SP Bel-Art Powder Funnels, Polypropylene, Bel-Art Products, a part of SP
Funnels Powder Funnels
Specially designed for transferring powders, these funnels have a wide stem for clog-free filling.
  • Polypropylene plastic construction
  • Angled, wide-stemmed design
  • Suited for transferring powders without clogging
  • Autoclavable
Made of polypropylene plastic with a 60º angle, they are steam autoclavable at 121ºC (250ºF).