Complete Photochemical Reaction System, Ace Glass Incorporated
Supplier: Ace Glass
7840-175 7840-180 7840-185 7840-360 7840-320 7840-340
89060-556EA 18266.13 CAD
89060-556 89060-558 89060-560 89084-402 89084-406 89084-410
Complete Photochemical Reaction System, Ace Glass Incorporated
Laboratory Reactor Systems
Complete reaction assembly with all parts needed for immediate operation
Borosilicate glass reactor has a standard taper 60/40 center joint, one- stadnard taper 14/20 angled joint for sparger tube, one-standard taper 24/40 vertical joint for condenser, and one-#7 Ace-Thred joint to accommodate thermometer. Volumes indicated are total volumes. Volume in reactive area of lamp is 40-50% of the total volume. Complete assembly consists of reactor, quartz immersion well, sparger tube, PTFE stir bar #7 thermometer bushing, three meters of 4.8 mm (3/16") I.D. PTFE tubing, stand, 450 watt lamp andMore Product Information