Photochemical Reaction Vessel, Ace Glass Incorporated
Supplier: Ace Glass
7841-04 7841-05 7841-06 7841-09 7841-10 7841-16 7841-03 7841-14 7841-15 7841-19
89060-564EA 1521.91 CAD
89060-564 89060-566 89060-568 89060-570 89060-572 89060-578 89061-362 89061-368 89061-370 89061-372
Photochemical Reaction Vessel, Ace Glass Incorporated
Reaction Vessels Reaction Vessels, Photochemical
Reaction vessel constructed of borosilicate glass to accommodate the immersion wells
With one- standard taper 14/20 angled joint for sparger tube, one- standard taper 24/40 vertical joint for condenser and one- #7 Ace-Thred side arm for thermometer. Center joint is standard taper 60/40. Bottom of reactor is flat to allow use of magnetic stirrer. Volumes indicated are total volumes. Volume in reactive area of lamp is approx. 40-50% of total volume.
Ordering information: Complete unit consists of vessel, sparger tube, PTFE stir bar, threaded nylon bushing and three meters of 4.8 mm (3/16"More Product Information