Photochemical Jacketed Reaction Vessel with Stopcock Bottom, Ace Glass Incorporated
Supplier: Ace Glass
7844-03 7844-06 7844-09
89060-582EA 2954.03 CAD
89060-582 89060-584 89060-586
Photochemical Jacketed Reaction Vessel with Stopcock Bottom, Ace Glass Incorporated
Reaction Vessels Reaction Vessels, Photochemical
Jacketed reaction vessel to enable cooling of reactant material during photolysis plus the added feature of a 2 mm bore, 1:5 PTFE stopcock at bottom for draining inner vessel contents
Constructed of borosilicate glass to accommodate immersion wells. With one- standard taper 14/20 angled joint for sparger tube, one- standard taper 24/40 vertical joint for condenser and one-#7 Ace-Thred side arm for thermometer. Center joint is standard taper 60/40. Bottom of reactor is flat to allow use of stir bar. Volumes indicated are total volumes. Volume in reactive area of lamp is approximately 40-50%More Product Information