SP Bel-Art ProCulture Foil Cover-ALLS, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a part of SP
F13200-1035 F13200-1055 F13200-1045
89087-684CS 708.5 CAD
89087-684 89087-688 89087-686
SP Bel-Art ProCulture Foil Cover-ALLS, Bel-Art Products, a part of SP
Aluminium Foils
Used in cell culture to cover flasks while allowing proper oxygenation, these economical and disposable covers also provide dust-free storage of clean labware.
  • Provides a temporary, sterile boundary for autoclave sterilization of labware
  • Preformed to quickly cover a wide variety of labware using only one hand
Great for use in centralized washing areas where cleaned labware needs to be transported back to the lab while remaining clean and contaminant-free.