Flo-Thru Reactor, Ace Glass Incorporated
Supplier: Ace Glass
9841-18 9841-30
89075-542EA 833.16 CAD
89075-542 89075-544
Flo-Thru Reactor, Ace Glass Incorporated
Reaction Vessels Reaction Vessels, Cylindrical Shape
Continuous-flow borosilicate glass vessel provides uniform treatment by forcing reactant to pass in front of horn tip
Must be used with 1/2" O.D. Ultrasonic horn and either 1/2 x 5" Extender or 1/4 x 5" Extender Microtip. Inlet and outlet tubes are 1/2" O.D. (13 mm). #7 Ace-Thred located below top thread is for bleed or vacuum connection. For latter use, bushing with hole needs to be ordered for tubulature connection. Operated in vertical position only.
Complete item consists of reactor, #7 PTFE plug and #25 nylon bushing. Slide adapter and horn sold separately.
The degree of processing is controlled by varying the power level and flow rate, max. 1.5L/Min. Reactants are pumped through side port, overflowing inner cup and out through bottom port. Treated material drains completely (no hang-up). The use of #36 slide adapter in #25 Ace-Thred at top allows probe position to be varied within the inlet cup area, thereby maximizing use of ultrasonic energy.