Ultrasonic Reaction Vessel, Small Volume, 3 to 10 ml, Ace Glass Incorporated
Supplier: Ace Glass
9843-25 9843-04
89075-546EA 710.77 CAD
89075-546 89075-548
Ultrasonic Reaction Vessel, Small Volume, 3 to 10 ml, Ace Glass Incorporated
Reaction Vessels Reaction Vessels, Cylindrical Shape
Tapered walls and proper size horn allow volumes as little as 3 mL to be mixed
Fabricated of borosilicate glass with #25 Ace-Thred center neck and two standard taper 14/20 side necks. With #25 bushing, center neck will accept #36 slide adapter in 25 mm with a horn and extender. Vessel measures 123 mm (4 7/8") high. Not supplied with bushing.