Reaction Assembly, Photochemical, Ace Glass Incorporated
Supplier: Ace Glass
7861-250 7861-255 7861-450 7861-410 7861-430
89061-358EA 17479.73 CAD
89061-358 89061-360 89084-404 89084-408 89084-412
Reaction Assembly, Photochemical, Ace Glass Incorporated
Reaction Vessels Reaction Vessels, Photochemical
Complete reaction assembly with all parts needed for immediate operation
Utilizes an internally threaded connection in place of the ground glass joint. Bushing and FETFE O-Ring form a compression type seal with immersion well. Well has removable inner cooling tube. Reactor has one- standard taper 14/20 angled joint for sparger tube, one- standard taper 24/40 vertical joint for condenser, and one- #7 Ace-Thred for thermometer. Volume indicated is total volume. Actual working volume is approximately 40-50% of total.
Ordering information: Complete assembly consists of reactor, 89061-380 quartz immersion well, sparger tube, PTFE Stir Bar, #7 nylon bushing with O-Ring, 3 Meters of 4.8 mm I.D. PTFE Tubing, stand, and 450 watt lamp and power supply.