Flexible Shaft Coupling, 28 mm, Ace Glass Incorporated
Supplier: Ace Glass
6472-155 6472-156 6472-159
89054-598EA 194.61 CAD
89054-598 89054-600 89083-348
Flexible Shaft Coupling, 28 mm, Ace Glass Incorporated
Stirrers Stirring Shafts
This is a coated steel flex-beam connector, for attaching directly to stir motor drive shaft.
Fits three different standard motor shaft sizes, top. Comes with 1/2" stainless steel pin, (bottom) that attaches to top of the 28 mm nylon chuck. Stir shaft diameter, 28 mm.