Duran Bottle GL 45 Connection System, Ace Glass Incorporated
Supplier: Ace Glass
7632-07 7632-06 7632-05 7632-01 7632-02 7632-10 7632-11 7632-12 7632-13
89214-900EA 176.8 CAD
89214-900 89214-902 89214-904 89214-912 89214-914 89214-930 89214-936 89214-940 89214-944
Duran Bottle GL 45 Connection System, Ace Glass Incorporated
Closures Bottle Closures
The heart of the GL45 connection system is a GL45 polypropylene cap with GL18 threaded ports
Modular accessories include polypropylene tube connection caps,solid polybutylene terepthalate (PBT) caps, PBT pressure equalization caps and PTFE cap inserts to accomodate various sizes of tubing. A typical application is the safe transfer of liquid media within a closed and sterile system for which you'd need one two-pack GL45 cap, two 89214-900 equalization caps, and some combination of solid caps and tubing connections with inserts.