Photochemical Cell Windows, Ace Glass Incorporated
Supplier: Ace Glass
7895-03 7895-08
89061-436EA 76.5 CAD
89061-436 89061-438
Photochemical Cell Windows, Ace Glass Incorporated
Reaction Vessels Reaction Vessels, Photochemical
Windows to be used with photochemical cells and cell window holder.
The windows are ground and polished (optical grade).
Order Now
SPECIFICATIONS
Environmentally Preferable