Sprayer, Clean-In-Place, Ace Glass Incorporated
Supplier: Ace Glass
6472-200 6472-201 6472-204 6472-206 6472-208 6472-210
89054-640EA 2402.98 CAD
89054-640 89054-642 89054-644 89054-646 89054-648 89054-650
Sprayer, Clean-In-Place, Ace Glass Incorporated
Atomizers
This sprayer is designed for cleaning ACE reactors without dismantling, or Clean-In-Place (CIP).
One moving part, a rotating disc at end of extension, produces heavy droplets in a 39" diameter, 360° pattern to simultaneously cover entire surface of reactor. Rotating disc is constructed of carbon-filled PTFE and stainless steel. Wash fluid enters through a series of holes onto a precision shaped disc. The disc rotates freely at high speed and the wash fluid is distributed in a dense spray. Can be operated in any position. Working pressure is 15 to 60 psi; maximum temperature is 120 °C. Fits through openings as small as 11/8" diameter, including a standard taper 45/50 outer joint. Assembly consists of spray disc, stainless steel coupling, extension tube, elbow, nipple, and a standard taper 45/50 inner to #15 Ace-Thred glass adapter with a nylon bushing and FETFE o-ring.