Ultrasonic Slide Adapter, Ace Glass Incorporated
Supplier: Ace Glass
9852-21 9852-25 9852-41 9852-45
89075-566EA 243.96 CAD
89075-566 89075-568 89075-570 89075-572
Ultrasonic Slide Adapter, Ace Glass Incorporated
Fittings and Connectors
This adapter is for use with a 1/2" ultrasonic horn threaded tip and 1/4" or 1/2" extenders, only
Complete item consists of adapter, one nylon bushing with one FETFE o-ring.
Both slide adapters have a #36 ace-thred at top with a 6" extension, either 25 mm O.D. for insertion into a #25 ace-thred, or 35 mm O.D. for use in a #36 ace-thred. Secure 1/2" horn in adapter with a bushing and o-ring, then slide adapter extension into thread on reaction vessel, again securing with a bushing. Now you have a variable depth adjustment of horn to achieve greater efficiency.