VWR® Standard Line Screw-Cap Microcentrifuge Tubes, PP
Supplier: VWR International
10025-742 10025-744 10025-746 10025-748 10025-750 10025-752 10025-754 10025-756 10026-096 10026-098 10026-102
10025-742CS 880.16 CAD
10025-742 10025-744 10025-746 10025-748 10025-750 10025-752 10025-754 10025-756 10026-096 10026-098 10026-102
VWR® Standard Line Screw-Cap Microcentrifuge Tubes, PP
Tubes Microcentrifuge Tubes
Crystal-clear polypropylene tubes are ideal for sample processing applications.
  • Available in three volumes: 0.5, 1.5, and 2.0 ml
  • Available in conical bottom and self-standing bottom formats
  • Tubes can be spun up to 20000×g
  • Caps and tubes are steam autoclavable at 121 °C and freezable to −80 °C
  • DNase-free, RNase-free, and non-pyrogenic
Excellent for either freezer storage or secure sample handling. Screw caps incorporate silicone O-Rings for a leakproof seal and accept color-coded inserts. Inserts snap into the top of the cap and create a flush surface. Caps and tubes are steam autoclavable. Sterile tubes are sterilized with irradiation.
Certifications: TSE/BSE-free Certification.
Ordering information: Bagged tubes are packaged in durable and re-sealable (zip closure) bags. Inner packaging is individually labeled for lot-to-lot traceability. Sterile tubes are sold with cap attached, non sterile tubes are sold without cap. Caps and color inserts are available as optional accessories and are sold separately.