VWR® Gravity Convection Ovens
Supplier: VWR International
89511-404 89511-406 89511-408 10144-898 10144-900 10144-902 10145-412 10145-414 10145-416
89511-404EA 4604.17 CAD
89511-404 89511-406 89511-408 10144-898 10144-900 10144-902 10145-412 10145-414 10145-416
VWR® Gravity Convection Ovens
Ovens Standard Ovens
VWR® Gravity Convection ovens are the optimal choice for heating and drying of delicate samples that require a gentle airflow.
  • Consistency - Repeatable results with outstanding temperature stability and uniformity
  • Versatility - Covers wide range of heating and drying applications
  • Extremely versatile with a wide temperature range, 50 – 250 °C and three benchtop sizes ranging from 2.3 – 6.2 cu. ft.
  • 2 wire mesh shelves included (55 lbs. capacity each)
  • Two-Year Warranty with Extended Warranty Packages Available
Powders and light samples will not be blown away by active air movement.
Control and display; the PID control is easy to use, and has a large display for optimal feeback to the operator. Timer function allows to turn oven on and off at desired time. Overtemperature alarm and a dual sensor safety concept are providing peace of mind. For data monitoring the oven can be connected with a standard RS-232 interface to a computer.
Chamber; cleaning is simple, due to stainless steel interior with smooth, rounded corners. The flexible shelving system with 13 to 19 positions allows taking best advantage of the internal volume. Heat stays inside the unit due to a unique insulation design, with benefit of low energy consumption and cool outside. Bench saving footprint does not compromise volume.
Processes such as sterilization or heated storage are followed economically. Air exchange can be controlled by a damper - open it for drying to remove humidity quickly, and close it for heat treatment with highest requirements for temperature accuracy.
Certifications: cCSAus certified.
Ordering information: Two-year standard warranty in US and Canada. For fast drying processes and highest temperature accuracy, please check VWR® Forced Air ovens.