Qualitative Filter Paper, Grade 617, Ahlstrom
Supplier: Ahlstrom
6170-1850 6170-3200 6170-0550 6170-1100 6170-2500 6170-1500 6170-5000 6170-0700 6170-0900 6170-2400 6170-4000 6170-3300 6170-0750 6170-1250
30617-442EA 198.43 CAD
30617-442 28342-032 47745-832 30617-440 47745-830 30617-438 75875-630 71440-006 47745-824 97058-892 47745-822 28342-028 97058-876 47745-826
Qualitative Filter Paper, Grade 617, Ahlstrom
Filters Filter Papers Qualitative Filter Paper
Grade 617 Qualitative Filter Papers are high-purity papers with high-speed filtration capabilities
They are made from 60% cotton and 40% wood fibers.