Samco™ Narrow Mouth Bio-Tite™ Specimen Containers, 90 ml, Thermo Scientific
Supplier: Samco
02 0148 02 0016 02 0039 02 0146 02 0214
15706-802CS 524.02 CAD
15706-802 89498-314 89498-318 89498-324 89498-334
Samco™ Narrow Mouth Bio-Tite™ Specimen Containers, 90 ml, Thermo Scientific
Sample Containers General Purpose Containers
Prevent bio-hazardous spills while conserving laboratory space with stackable Thermo Scientific™ Samco™ Narrow-Mouth Bio-Tite™ 90 mL (3 oz.), 48 mm specimen containers
These shatter-resistant containers feature a proprietary, all plastic, leak-resistant seal. They are tested at varying pressures, making them ideal for use in various transport systems. Containers are pre-assembled with caps attached.