iMiniPlus® Dry Ice Loggers, Cryopak Verification Technologies
Supplier: Cryopak Verification Technologies
MP-OE-N-8-L MP-ST-N-8-L
97017-230EA 1013.57 CAD
97017-230 10033-818
iMiniPlus® Dry Ice Loggers, Cryopak Verification Technologies
Data Loggers
The iMiniPlus® Dry Ice logger can be programmed to measure extreme temperatures within the range of -100°C to 40°C (-148°F to 104°F)
  • Start and stop buttons on the face of the unitx
  • Bookmarking capabilities
  • Large LCD display with multifunctional viewing
  • Large memory capacity up to 8040 readings
  • Polycarbonate housing
Devices have a 1 m cable that connects the stainless steel temperature probe to the data logger. Available as a single-trip or multi-use unit.
It is the ideal solution for ensuring safe transport and storage of perishables held in dryMore Product Information
Order Now
SPECIFICATIONS
Sensor Ext. sensor, T90 of 5 min. in moving air
Environmentally Preferable
Alarm High and low alarm configuration, 2 thresholds
Interface
Software Console Pro
Protection class IP 65
Battery life 1 - 2 years
Weight 70 g
W×D×H 83×57×18 mm
Data memory - No. of readings 8048
Batteries User resplaceable, lithium button cell (CR 2450), 3 V
Start delay Yes