Epoxy Adhesive, Ace Glass Incorporated
Supplier: Ace Glass
7560-10
89059-444EA 135.09 CAD
89059-444
Epoxy Adhesive, Ace Glass Incorporated
Adhesives
A two component epoxy system, which cures to a chemical resistant film.
  • Cures to a chemical resistant film
  • Bonds tightly
  • 30mL jar
The epoxy is used to tightly bond a PTFE sleeve to a glass joint. Supplied in 30 mL jars with the activator in a separate container. The 30 mL supply will cement at least 25 joints depending on size.