Joints, Socket, Spherical, Ace Glass Incorporated
Supplier: Ace Glass
7558-02 7558-04 7558-06 7558-08 7558-10
89059-434EA 140.5 CAD
89059-434 89059-436 89059-438 89059-440 89059-442
Joints, Socket, Spherical, Ace Glass Incorporated
NS Joint Sleeves and Connectors Joint Sleeves
Recommended for use with PTFE-clad ball member
Polished surface does not wear PTFE, assures more precise fit than the standard ground surface.