High Torque Economy Laboratory Stirrers, Ace Glass
Supplier: Ace Glass
13650-12 13650-23 13650-40
89077-824EA 1211.56 CAD
89077-824 89077-826 89077-828
High Torque Economy Laboratory Stirrers, Ace Glass
Stirrers Overhead Stirrers
Motor and controller, motor is non-reversible, 1/12 hp DC, 5/16" dia shaft 1" long
stand shown in image is sold separately.
Recommended for use with 8081 Flexible Shaft or general stirring. Motor is adjustable from 280 - 2800 rpm and generates 60 oz/inch of nominal torque. Motor can only be used with the 120 V, 50/60Hz controller and is supplied with a 5/8" diameter mounting rod
Ordering information: Ace Glass Laboratory Stirrers, High Torque, Economy Complete Package Includes (1) Each:
89077-824
89077-826
Order Now
SPECIFICATIONS
Environmentally Preferable