Hydrogenation/Gas Apparatus I, Ace Glass Incorporated
Supplier: Ace Glass
7482-20 7482-25 7482-31 7482-33 7482-37 7482-38 7482-42 7482-46 7482-50 7482-500
89058-884EA 17566.96 CAD
89058-884 89058-886 89058-888 89058-890 89058-892 89058-894 89058-896 89058-898 89058-900 89058-902 89058-906 10767-186
Hydrogenation/Gas Apparatus I, Ace Glass Incorporated
Reaction Vessels Reaction Vessels, Cylindrical Shape
Parr Model 3911 Hydrogenation unit for working volumes of 1 to 200 mL
Modified Vessel Clamp Allows Clamping of 89051-324 Temperature Measurement Adapter to Parr Model 3911 Hydrogenation Units
Nylon Adapter Connects Vessel to Gas Tank Tubing On Smaller #7 Ace-Thred Vessels
PTFE Adapter Connects Vessel to Gas Tank Tubing On The Larger #25 Ace-Thred Vessels
Upper and Lower Bumper Pad Protects Glassware In 89058-884 and 89058-908 Hydrogenation Units
Ace-thred vessels replace rubber stoppered vessels and eliminate cross-over contamination caused by leeching or sample retension. Complete unit includes all adapters needed for bottles (1 - 4 mL, 3 - 8 mL, 8 - 20 mL, 10 - 35 mL, 25 - 75 mL and 50 - 200 mL). Units without vessels and adapters are available in explosion proof and air driven models.
Ordering information: No glassware or adapters included, except for the complete unit (7482-50).