iMini® Temperature Logger, Cryopak Verification Technologies
Supplier: Cryopak Verification Technologies
MX-ST-S-8-L MX-IN-S-8-L MX-OE-S-8-L MX-1E-S-8-L MX-2E-S-8-L MX-HS-S-16-L
10033-794EA 110.65 CAD
10033-794 10033-796 10033-798 10033-800 10033-802 10033-804
iMini® Temperature Logger, Cryopak Verification Technologies
Data Loggers
The iMini® series, provided by Cryopak Verification Technologies, is available as a single trip or multi-use data logger
  • 24 months warrenty, excluding battery (Multi use loggers), One Trip, excluding battery (Single trip loggers)
  • Password protected, (programmable using ConsolePlus Software) encrypted binary file is generated along with PDF
  • The multifunctional LCD screen then provides several levels of information:
  • The highest temperature, the lowest temperature and the average temperature recorded
  • Any alarms triggered
  • The time spent outside alarm ranges
  • An icon if the battery is depleted
The iMini® Temperature Logger series was developed to encompass a full spectrum of uses, from food processing and warehousing, to blood storage and transport, to biotech clinical trials and beyond. The device can be programmed in either Celsius or Fahrenheit, and downloaded using a USB nonproprietary interface cable.
All Temperature Loggers offer an IP rating of IP 65.
Standard features include a four alarm threshold with LED indicators, large LCD visibility, and start/stop buttons. Enhanced options include NIST traceable calibration and bookmarking capability. This series offers full versatility of memory capacity and sensor array. With five memory options from 8000 to 128,000 readings and four sensor configurations, these temperature recorders adapt to a multitude of industry specific applications.