Borosilicate Glass Beaded Pipe Adapter, Ace Glass Incorporated
Supplier: Ace Glass
6472-277 6472-278 6472-276 6472-275
89082-542EA 208.72 CAD
89082-542 89082-544 89082-546 89082-548
Borosilicate Glass Beaded Pipe Adapter, Ace Glass Incorporated
Fittings and Connectors
Borosilicate glass beaded pipe adapter
Used as a transition adapter on flush seal drain valves with 1" beaded pipe side arms.