Microscale Photochemical Reaction Assembly, Ace Glass Incorporated
Supplier: Ace Glass
7880-12 7880-15 7880-20 7880-60 7880-62
89061-390EA 212.37 CAD
89061-390 89061-392 89061-394 89061-396 89061-398
Microscale Photochemical Reaction Assembly, Ace Glass Incorporated
Reaction Vessels Reaction Vessels, Photochemical
Reactor body is fabricated of borosilicate glass with #25 Ace-Thred at top, standard taper 14/10 outer side joint externally threaded to allow an O-Ring cap seal connection with mating inner joint, and porosity E (4-8micron) filter disc at bottom for use as a gas inlet to remove oxygen from the solution and/or for agitation (mixing)
Bottom hose connection is size E. Immersion well is either quartz or borosilicate, 11 mm I.D., with water jacket. Well can also be used to flow filter solutions in front of lamp to attain desired wavelengths. Well is assembled by insertion into #25 Ace-ThredMore Product Information