Photochemical Quartz Immersion Well, Low Temperature, Ace Glass Incorporated
Supplier: Ace Glass
7876-10 7876-50
89084-932EA 5811.18 CAD
89084-932 89084-930
Photochemical Quartz Immersion Well, Low Temperature, Ace Glass Incorporated
Reaction Vessels Reaction Vessels, Photochemical
Triple walled, quartz, immersion well for use in temperatures as low as –78 °C
Same as the immersion well, except, plain-sided, (without standard taper joint), for use with #50 Ace-Thred bushing and all vessels. 415 mm total jacket length. Will accommodate 89061-252 and 89061-256 inner tubes. Fabricated from borosilicate glass.