Unjacketed Cylindrical Reaction Flask, KF Plane Flange, Ace Glass Incorporated
Supplier: Ace Glass
6472-241 6472-242 6472-248 6472-249 6472-64 6472-65 6473-05 6473-11
89054-656EA 8559.92 CAD
89054-656 89054-658 89054-662 89054-664 89054-766 89054-768 89054-808 89054-818
Unjacketed Cylindrical Reaction Flask, KF Plane Flange, Ace Glass Incorporated
Reaction Vessels Reaction Vessels, Cylindrical Shape
These unjacketed cylindrical reaction flask are made of heavy wall borosilicate glass and include a bottom outlet valve
Reactor accessories including Flat Flange Couplings, Flat Flange PTFE Gaskets, Bottom Outlet Valves, Beaded Pipe Couplings for Valves, Spherical Joint Adapters for Valves, and Spherical Joint O-Rings are sold separately.
Each flask has high visibility capacity reference graduations for easy viewing. They can be used standalone or with bottom opening heating mantles. Available with Schott-Duran O-ring groove or KF Plane flat ground flanges. as well as a variety ofMore Product Information