Gripper Fitting Connector With Sealing Plug, Ace Glass Incorporated
Supplier: Ace Glass
12724-04 12724-11 12724-12 12724-15 12724-16 12724-25
89077-446EA 74.75 CAD
89077-446 89077-448 89077-450 89077-452 89077-454 89077-456
Gripper Fitting Connector With Sealing Plug, Ace Glass Incorporated
Fittings and Connectors
This high pressue tubing connector can be used with PTFE or plastic tubing with 1.6 mm outer diameter
It is rated at 1000 psi, and has an 1/4"-28 thread end fitting. Gripper fitting is made of high tolerance 316 S.S. machined on precision Swiss lathes and is for use with PTFE or plastic tubing having 1/16 or 1/8" O.D. The sealing plug consists of Tefzel body and lock nut, PTFE sealing face, and is for use with 1/4"-28 female threaded fittings.