Laboratory Stirrers, Ace Glass Incorporated
Supplier: Ace Glass
13649-07 13649-09 13649-19
89077-804EA 1910.57 CAD
89077-804 89077-806 89077-808
Laboratory Stirrers, Ace Glass Incorporated
Stirrers Overhead Stirrers
Totally enclosed, 1/40hp DC permanent magnet motor with dual 8 mm (5/16") armature and gear shafts with milled flats
Motor is supplied with 1.5 meter three-wire cable with plug and ground lead for connection to controller. Measures: 5-1/2" x 4" x 4". Weight: 4 lbs. 7 oz. Controller is solid state 120 V, 10 amp AC maximum or 0–120 V, 6 amp DC maximum. Features rear ring stand clamp, Forward-Off-Reverse switch. Supplied with heavy duty 1.8 meter three-wire power cord with NEMA plug. Supplied with one-amp fuse to protect motor. Controller measures 102 (4") x 54 mm (2-1/8") x 41 mm (1-5/8").More Product Information