Micro/Mini-Lab Kit, Deluxe Kits, Ace Glass Incorporated
Supplier: Ace Glass
9560-30 9560-05 9560-14 9560-18 9560-62 9560-63 9560-66
71001-658EA 1742.93 CAD
71001-658 89075-124 89075-126 89075-128 89075-132 89075-138 89075-142
Micro/Mini-Lab Kit, Deluxe Kits, Ace Glass Incorporated
Labware Sets
This is a Deluxe Micro/Mini-Lab Kit for performing all of the experiments in Organic Laboratory Techniques.
  • Kits come with a variety of components
  • Includes vials, condensers, tubes, columns, syringes, plugs and more
  • Supplied in compartmentalized or foam insert plastic tilt lid boxes
The basic kit will do the majority of experiments in Organic Laboratory Techniques, but this deluxe kit will do all experiments. Kits come with variety of components such as vials, condensers, tubes, the new Hickman-Hinkle spinning distillation column, retrieval syringeMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable