Volumetric Bulbs, Ace Glass Incorporated
Supplier: Ace Glass
8004-03 8004-10 8004-15 8004-18 8004-21 8004-22 8004-23 8004-24 8004-27 8004-30 8004-33 8004-36 8004-39 8004-42 8004-44 8004-45
89061-590PK 39.31 CAD
89061-590 89061-592 89061-594 89061-596 89061-598 89061-600 89061-602 89061-604 89061-606 89061-608 89061-610 89061-612 89061-614 89061-616 89061-618 89061-620
Volumetric Bulbs, Ace Glass Incorporated
Bottle Top Tilt Measures
Fills and pours automatically, rapidly.
  • One head for all capacities, all volumes
  • Short tipping angle, interchangeable volumetric bulbs give reproducible volumes
Bulb joint is a standard taper 14/20, head and flask standard taper 29/42.
Complete apparatus consists of head, flask, volumetric bulb, clip, two 19 mm (3/4") springs, one 38 mm (1½") spring.