Socket, Rubber Septum, Ace Glass Incorporated
Supplier: Ace Glass
7663-01 7663-02 7663-04
89060-222EA 27.68 CAD
89060-222 89060-224 89060-226
Socket, Rubber Septum, Ace Glass Incorporated
Adapters Adapters, Conical Ground Joints
The end of this socket is tooled to accept a rubber stopper septa.
  • Accepts rubber stopper septa
  • Use on chromatography columns
This socket is used on chromatography columns. Overall length is 15 cm and opening is 5 mm.
Order Now
SPECIFICATIONS
Environmentally Preferable