Micro/Mini-Lab Stainless Steel Gow-Mac GC Adapter, Ace Glass Incorporated
Supplier: Ace Glass
9571-40 9571-42
89075-172EA 225.99 CAD
89075-172 89075-174
Micro/Mini-Lab Stainless Steel Gow-Mac GC Adapter, Ace Glass Incorporated
Fittings and Connectors
Microscale gas chromatography connection-adapter for making direct connection between GC column and 89075-192 collection tube.
  • Stainless steel
  • With NPT female thread and externally threaded 5/5 outer joint
  • Allows “O-Ring- CAP-SEAL” connection via a tube
Stainless steel adapter has a 6-32 NPT female thread for attaching to heated port on GC and a 5/5 outer joint, externally threaded, to allow an “O-Ring- CAP-SEAL” connection via a tube.