Sublimation Adapter, Ace Glass Incorporated
Supplier: Ace Glass
8015-15 8015-20
89061-622EA 567.62 CAD
89061-622 89061-624
Sublimation Adapter, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
Unique device for carrying out sublimation directly in a round bottom reaction flask.
  • Coolant connection at top
  • Side vacuum connection
  • 1:5 PTFE 2 mm bore stopcock
  • [ST] joint for use with 250 mL or 500 mL flasks
Simply remove condenser, stirrer, etc. from flask joint, insert sublimation adapter and begin. Coolant connection at top and side vacuum connection are 9.5 mm (3/8") O.D. Vacuum connection is controlled by a 1:5 PTFE 2 mm bore stopcock. Adapter is supplied with a standard taper 24/40 joint for use with 250 mL or 500 mL flasks. Other joint sizesMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable