Micro/Mini-Lab Spinning Band Distilling Column, Ace Glass Incorporated
Supplier: Ace Glass
9595-05 9595-07 9595-43 9595-47
89075-296EA 493.86 CAD
89075-296 89075-298 89075-300 89075-302
Micro/Mini-Lab Spinning Band Distilling Column, Ace Glass Incorporated
Distillation Components Distillation Sets
High performance, low cost, Microscale Spinning Band Distillation Column system effective in separating low-boiling liquid mixtures with volumes in the 0.5 to 5 mL range.
  • Separates close-boiling mixtures
  • Achieves height equivalent/theoretical plate values approaching 0.2"/plate within 90 minutes of boil-up
  • Utilizes PTFE band uniquely driven by a bottom magnet
Designed by Drs
The column top joint is a combination cap, thread and standard taper joint to accommodate and seal other microscale apparatus. The O.D. of the top joint has an external threadMore Product Information