Vacuum Sublimation Apparatus, Improved, Ace Glass Incorporated
Supplier: Ace Glass
8023-20 8023-25 8023-33 8023-36 8023-40 8023-55
89061-632EA 474.12 CAD
89061-632 89061-634 89061-636 89061-638 89061-640 89061-642
Vacuum Sublimation Apparatus, Improved, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
Available in two sizes, both featuring an o-ring sealed, greaseless flange with a quick release clamp.
  • Two sizes available
  • O-ring sealed, greaseless flange
  • With quick release clamp
The top flange is ground flat. The bottom mating flange has an O-Ring groove for a CAPFE O-Ring (PTFE encapsulated silicone), held tightly together via a quick release clamp. Size 60 is 200 mm high x 60 mm I.D.; uses 89055-106 clamp and CAPFE O-Ring. Size 152 is 250 mm high x 152 mm I.D.; uses 89055-112 clamp and CAPFE O-Ring. Complete item consists of outer body, inner top, CAPFE O-Ring and quick release clamp.