TruCLEAN® Cleanroom and Microfiber Mops, Perfex
Supplier: Perfex
22-32 22-29 22-31
89096-036CS 432.29 CAD
89096-036 89096-038 89096-040
TruCLEAN® Cleanroom and Microfiber Mops, Perfex
Mops
TruCLEAN Cleanroom Mop is constructed of continuously woven polyester fabric, individually packaged and laundered in a class 100 cleanroom facility
TruCLEAN Microfiber Mop contains many strands of synthetic fiber made of polyester and polyamide for superior cleaning capabilities.
TruCLEAN Anti-Microbial Microfiber Mop has an anti-microbial agent permanently bonded to the TruCLEAN microfiber, simultaneously killing microorganisms while inhibiting bacteria growth on surfaces.
Certifications: Compatible with gamma, ETO and autoclave sterilization – up to 250º F (121º C) for 30 minutes.More Product Information