Nalgene® Pharma-Grade Platinum-Cured Silicone Tubing, Thermo Scientific
Supplier: Thermo Fisher Scientific
8600-3173 8600-3182 8600-0020 8600-0030 8600-0040 8600-0050 8600-0060 8600-0070 8600-0080 8600-0100 8600-0120 8600-0130 8600-0140 8600-0190 8600-0200 8600-0260 8600-0310 8600-3015 8600-3016 8600-3017 8600-3018 8600-3024 8600-3025
89493-604CS 609.82 CAD
89493-604 89493-606 89501-666 89501-668 89501-670 89501-672 89501-674 89501-676 89501-678 89501-680 89501-682 89501-684 89501-686 89501-688 89501-690 89501-692 89501-694 89501-696 89501-698 89501-700 89501-702 89501-704 89501-706
Nalgene® Pharma-Grade Platinum-Cured Silicone Tubing, Thermo Scientific
Tubing Transfer Tubing
Nalgene® Pharma-grade Platinum-cured silicone tubing is flexible, durable, and translucent.
  • High-purity tubing
  • Excellent elastic memory
  • Ideal for a variety of transfer applications such as pharmaceutical, laboratory, and bioprocess manufacturing
Resiliency coupled with low hysteresis and compression set ensure that fluid transfers are reliable and accurate every time.
To fit in a wide range of applications, the new Nalgene® Pharma-Grade Platinum-cured silicone tubing can be sterilized by most methods, including autoclaving, gamma-irradiation, andMore Product Information
Order Now
SPECIFICATIONS
Model 89493-604 89493-606 89501-666 89501-668 89501-670 89501-672 89501-674 89501-676 89501-678 89501-680 89501-682 89501-684 89501-686 89501-688 89501-690 89501-692 89501-694 89501-696 89501-698 89501-700 89501-702 89501-704 89501-706
Environmentally Preferable
Max. pressure 8 psi 8 psi 18 psi 18 psi 14 psi 18 psi 12 psi 15 psi 18 psi 13 psi 9 psi 12 psi 14 psi 10 psi 12 psi 10 psi 10 psi 11 psi 11 psi 7 psi 6 psi 9 psi 9 psi