Pocket Pro Tester, Temperature, Hach
Supplier: Hach
9531700 9531701
89508-370EA 637.2 CAD
89508-370 89508-372 CA89508-370 CA89508-372
Pocket Pro Tester, Temperature, Hach
Thermometers Digital Thermometers
Hach Pocket Pro and Pro+ are engineered to deliver accurate results.
  • Waterproof (IP 67 and floats)
  • 1 point calibration (customer-defined)
  • Selectable Temp (°C or °F)
  • Auto-Off; Reading Hold
  • Quick-Release Lanyard (included)
  • Transparent Sensor Cap
  • Confidence in results: To maximize accuracy all testers are factory calibrated
Save time and money; easy-to-replace AAA batteries and convenient sample cup save time on maintenance and measurements. Ease of use; intuitive user interface guides users through the calibration and measurement process with a large, easy-to-read LCD screen. No ATC or Calibration.