SP Bel-Art Swing Jaw™ Tubing Clamps, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
H18220-0000 H18222-0000
70690-120PK 164.45 CAD
70690-120 70690-124
SP Bel-Art Swing Jaw™ Tubing Clamps, Bel-Art Products, a part of SP
Clamps Tubing and Sanitary Clamps
Zinc-plated steel tube clamp provides a non-slip closure for all flexible tubing – even heavy-walled vacuum lines.
  • Zinc-plated steel tube clamp construction
  • Can be installed without disconnecting tubing
  • Smooth clamping surfaces
A knurled nut compresses against a swinging jaw with smooth clamping surfaces. Can be installed and removed without disconnecting tubing.