VWR® Basic Reach-In Refrigerators with Glass Doors
Supplier: VWR
VWR®
TS72G-CHR TS23G-CHR TS49G-CHR
89239-070EA 21991.39 CAD
89239-070 89239-074 89239-072
VWR® Basic Reach-In Refrigerators with Glass Doors
Refrigerators
Traditional reach-in glass door refrigerators are available in single, double, or triple door formats.
  • Stainless steel siding
  • Heavy-duty shelving
  • Available in a variety of door formats
Each unit features a stainless steel front exterior and sides, with aluminum back and stainless steel interior
The 651 L capacity refrigerator also features one access port, one duplex outlet, and factory installed electronic temperature controller (in °C) with two probes and a digital display. The 1388 and the 2039 L capacity refrigerators also feature two access ports, one duplex outlet, and factory installed electronic temperature controller (in °C) with two probes and a digital display.
Units are equipped with energy efficient low E, thermal insulated glass door(s) with solar digital temperature display. Refrigerators maintain an average temperature of 3-5°C. All units are equipped with adjustable, heavy-duty, PVC-coated wire shelves with shelf clips for installation. Additional standard features include 10.2 cm (4") diameter swivel castors and door locks.
Certifications: UL Listed. NSF certified. CE marked.
Ordering information: Refrigerators come with a three-year parts and labor warranty, with an additional two-year warranty on the compressors.
Caution: Note: Dimensions shown below are external W×D×H.