Sodium chloride ≥99% ACS, Millipore®

Supplier: MilliporeSigma

567440-1KG 567440-500GM
CA80058-640EA 192.21 CAD
CA80058-640 CA80058-642
Sodium chloride ≥99% ACS, Millipore®
Sodium chloride
Formula: NaCl
MDL Number: MFCD00003477
CAS Number: 7647-14-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR