Sodium chloride 0,9% aqueous solution Normal saline solution, sterile

Supplier: TEKNOVA CA

Synonyms: Saline, NaCl, Sea salt, Halite

S5820 S5812 S5815 S5819 S5825
CA100216-354EA 380.64 CAD
CA100216-354 CA101320-568 CA101320-570 CA101320-572 CA101320-574
Sodium chloride 0,9% aqueous solution Normal saline solution, sterile
Sodium chloride

Dionized Water used as a solvent or diluent for antibiotics and biologicals.


  • Rack of 25 tubes in a reusable rack
  • Stored at room temperature

Formula: NaCl
MW: 58.44 g/mol
Boiling Pt: 1413
Melting Pt: 801
MDL Number: MFCD00003477
CAS Number: 7647-14-5
Merck Index: 14,08599

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR