Formaldehyde (Paraformaldehyde) 16% in aqueous solution, EM Grade

Supplier: Electron Microscopy Sciences
Danger

Synonyms: Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde

15700 15710-S 15711 15710
100503-914EA 21.19 CAD
100503-914 100503-020 100504-782 100503-917
Formaldehyde (Paraformaldehyde) 16% in aqueous solution, EM Grade
Formaldehyde

Paraformaldehyde - methanol free solution.


  • Methanol free
  • Microfiltered and stored in pre-scored ampoules


A more efficient and rapid penetrant fixative used in combination with glutaraldehyde and acrolein fixatives.


Easy to break, prescored, 10 ml ampoules sealed under inert gas, or 100 ml bottles.

Formula: CH₂O
Boiling Pt: >93 °C (>199 °F)
Density: 0.9392 g/cm³ (7.838 lbs/gal.)
Storage Temperature: Ambient
MDL Number: MFCD00003274
CAS Number: 50-00-0
UN: 2209
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f240682c03df85e943acbb1a983fe71750db44cd&vid=96c37abe2cf4c6160dda3947fb05d9d23c32fa74&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR