(3-Fluoro-4-methoxyphenyl)acetic acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.43872.0005 8.43872.0010
CA8.43872.0005CS 187158 CAD
CA8.43872.0005 CA8.43872.0010
(3-Fluoro-4-methoxyphenyl)acetic acid for synthesis, Sigma-Aldrich®
(3-Fluoro-4-methoxyphenyl)acetic acid
Formula: C₉H₉FO₃
MDL Number: MFCD00060353
CAS Number: 452-14-2

Order Now

Specification Test Results

Molar Mass 184.16 g/mol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR