Triisopropyl borate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: Tri-i-propylborate

M-Clarity™ quality level = MQ200

8.41264.0050 8.41264.0100 8.41264.0500
CA8.41264.0050CS 9081 CAD
CA8.41264.0050 CA8.41264.0100 CA8.41264.0500
Triisopropyl borate for synthesis, Sigma-Aldrich®
Triisopropyl borate
Formula: C₉H₂₁BO₃
Boiling Pt: 139 °C (1013 hPa)
Melting Pt: –59 °C
Density: 0.82 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008872
CAS Number: 5419-55-6
EINECS: 226-529-9
UN: 2616
ADR: 3,II

Order Now

Specification Test Results

Melting Point -59 °C
Molar Mass 188.07 g/mol
Boiling Point 139 - 141 °C (1013 hPa)
Vapor Pressure 75 hPa (100 °C)
Flash Point 25.5 °C
Density 0.82 g/cm3 (20 °C)
Hazard Class 3
Hazard Class Description Flammable liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR