Isopropyl myristate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Tetradecanoic acid isopropyl ester, Myristic acid isopropyl ester, Isopropyl Myristate, Isopropyl tetradecanoate, Tetradecylic acid isopropyl ester

M-Clarity™ quality level = MQ200

8.22102.0100 8.22102.1000 8.22102.2500
CA8.22102.0100CS 3257 CAD
CA8.22102.0100 CA8.22102.1000 CA8.22102.2500
Isopropyl myristate for synthesis, Sigma-Aldrich®
Isopropyl myristate
Formula: C₁₇H₃₄O₂
Boiling Pt: 320 °C (1013 hPa)
Melting Pt: 0…1 °C
Density: 0.852 g/cm³ (20 °C)
Flash Pt: 150 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008982
CAS Number: 110-27-0
EINECS: 203-751-4
Merck Index: 13,05235

Order Now

Specification Test Results

Solubility  - 0.05 mg/l (20 °C)
Molar Mass 270.45 g/mol
Boiling Point 140 °C (3 hPa)
Vapor Pressure  - 1 hPa (20 °C)
Flash Point 150 °C
Refractive Index 1.434 (20 °C, 589 nm)
Density 0.85 g/cm3 (20 °C)
Ignition Point 300 °C
Viscosity kinematic 6.57 mm2/s (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR