(±)-2-Methyltetrahydrofuran stabilized for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
M-Clarity™ quality level = MQ200

8.21093.0005 8.21093.0250 8.21093.1000
CA8.21093.0005CS 6020 CAD
CA8.21093.0005 CA8.21093.0250 CA8.21093.1000
(±)-2-Methyltetrahydrofuran stabilized for synthesis, Sigma-Aldrich®
(±)-2-Methyltetrahydrofuran
Formula: C₅H₁₀O
MDL Number: MFCD00005367
CAS Number: 96-47-9
UN: 2536
ADR: 3,II

Order Now

Specification Test Results

Solubility 150 g/l (25 °C)
Melting Point -136 °C
Molar Mass 86.13 g/mol
Boiling Point 78 - 80 °C (1013 hPa)
Vapor Pressure 136 hPa (20 °C)
Flash Point -11 °C
Refractive Index 1.4056 (20 °C, 589 nm)
Explosion Limit 1.2 - 5.7 %(V)
Density 0.86 g/cm3 (20 °C)
Ignition Point 270 °C
Hazard Class 3
Hazard Class Description Flammable liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR