4-(Dimethylamino)pyridine zS, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: 4-Dimethylaminopyridine, DMAP, N,N-Dimethylpyridin-4-amine

MQ200

8.20499.0005 8.20499.0100 8.20499.0250 8.20499.5000
CA8.20499.0005CS 74774 CAD
CA8.20499.0005 CA8.20499.0100 CA8.20499.0250 CA8.20499.5000
4-(Dimethylamino)pyridine zS, Sigma-Aldrich®
4-(Dimethylamino)pyridine
Formula: C₇H₁₀N₂
MW: 122.17 g/mol
Boiling Pt: 195
Flash Pt: 124
MDL Number: MFCD00006418
CAS Number: 1122-58-3
UN: 2811
ADR: 6.1 I

Order Now

Specification Test Results

Solubility 60 g/l (20 °C)
Melting Point 110 - 112 °C
Molar Mass 122.17 g/mol
Bulk Density 520 kg/m3
Boiling Point 138 - 140 °C (24 hPa)
Flash Point 110 °C
pH 11 (60 g/l, H2O, 20 °C)
Ignition Point 420 °C
Hazard Class 6.1
Hazard Class Description Toxic materials


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR