2-Bromethylamine hydrobromide for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
M-Clarity™ quality level = MQ200
8.20176.0100
8.20176.0500
8.20176.5000
CA8.20176.0100CS
7354
CAD
CA8.20176.0100
CA8.20176.0500
CA8.20176.5000
2-Bromethylamine hydrobromide for synthesis, Sigma-Aldrich®
Formula:
BrCH₂CH₂NH₃Br |
MDL Number:
MFCD00012886 CAS Number: 2576-47-8 |
Specification Test Results
Solubility |
|
Melting Point | 171 - 176 °C |
Molar Mass | 204.89 g/mol |
pH | 4.7 (25 g/l, H2O, 20 °C) |
Hazard Class | 6.1 |
Hazard Class Description | Toxic materials |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...